Assay Details
| Gene Symbol: Eps8l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133191.2 | NP_573454.2 | 11 - 12 | 1094 | 60 | |
| GenBank mRNA | BC005492.1 | - | 11 - 12 | 1094 | 3487337 | 60 |
| BC009098.1 | - | 11 - 12 | 1152 | 3482190 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eps8l2
Genomic Map