Assay Details
| Gene Symbol: 1700012A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC059718.1 | - | 2 - 3 | 181 | 72 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CA466124.1 | - | 184 | Chr. - - [-] on Build | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13848 | Gm13848 | OTTMUSG00000014743 | -385.2kb | Anti-sense | Chr. 6 - 31658173 - 31666131 on NCBI Build GRCm38 |
| Downstream | plexin A4 | Plxna4 |
9330117B14, Plxa4, mKIAA1550 |
85.6kb | Anti-sense | Chr. 6 - 32144543 - 32588192 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

1700012A03Rik
Genomic Map