Assay Details
| Gene Symbol: Fzd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162494.1 | NP_001155966.1 | 3 - 4 | 696 | 71 | |
| NM_008056.3 | NP_032082.3 | 3 - 4 | 700 | 71 | ||
| GenBank mRNA | AK088399.1 | - | 3 - 4 | 565 | 71 | |
| BC026150.1 | - | 3 - 4 | 689 | 3983985 | 71 | |
| U43319.1 | - | 2 - 3 | 526 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fzd6
Genomic Map