Assay Details
| Gene Symbol: Dcaf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198606.2 | NP_941008.2 | 7 - 8 | 898 | 104 | |
| GenBank mRNA | BC060375.1 | - | 7 - 8 | 840 | 5038466 | 104 |
| BQ572009.1 | - | 4 - 5 | 376 | 104 | ||
More Information
Set Membership: |
|

Dcaf13
Genomic Map