Assay Details
| Gene Symbol: Dcaf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198606.2 | NP_941008.2 | 9 - 10 | 1192 | 124 | |
| GenBank mRNA | AK139315.1 | - | 2 - 3 | 6127 | 124 | |
| AK157825.1 | - | 2 - 3 | 4447 | 124 | ||
| BB262409.2 | - | 2 - 3 | 197 | 124 | ||
| BC060375.1 | - | 9 - 10 | 1134 | 5038466 | 124 | |
| W42174.1 | - | 1 - 2 | 52 | 124 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dcaf13
Genomic Map