Assay Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029982.2 | NP_001025153.2 | 10 - 11 | 2012 | 115 | |
| GenBank mRNA | AK048053.1 | - | 10 - 11 | 1941 | 115 | |
| BC039781.1 | - | 2 - 3 | 196 | 115 | ||
| BC066195.1 | - | 9 - 10 | 1643 | 115 | ||
| BC098506.1 | - | 10 - 11 | 1901 | 30014037 | 115 | |
| BC116304.1 | - | 9 - 10 | 1393 | 115 | ||
| BC116305.1 | - | 9 - 10 | 1393 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec23ip
Genomic Map