Assay Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029982.2 | NP_001025153.2 | 13 - 14 | 2460 | 85 | |
| GenBank mRNA | BC039781.1 | - | 5 - 6 | 644 | 85 | |
| BC066195.1 | - | 12 - 13 | 2091 | 85 | ||
| BC098506.1 | - | 13 - 14 | 2349 | 30014037 | 85 | |
| BC116304.1 | - | 12 - 13 | 1841 | 85 | ||
| BC116305.1 | - | 12 - 13 | 1841 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec23ip
Genomic Map