Assay Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029982.2 | NP_001025153.2 | 14 - 15 | 2619 | 115 | |
| GenBank mRNA | BC023480.1 | - | 1 - 2 | 68 | 115 | |
| BC039781.1 | - | 6 - 7 | 803 | 115 | ||
| BC066195.1 | - | 13 - 14 | 2250 | 115 | ||
| BC098506.1 | - | 14 - 15 | 2508 | 30014037 | 115 | |
| BC116304.1 | - | 13 - 14 | 2000 | 115 | ||
| BC116305.1 | - | 13 - 14 | 2000 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec23ip
Genomic Map