Assay Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029982.2 | NP_001025153.2 | 9 - 10 | 1889 | 109 | |
| GenBank mRNA | AK048053.1 | - | 9 - 10 | 1818 | 109 | |
| BC039781.1 | - | 1 - 2 | 73 | 109 | ||
| BC066195.1 | - | 8 - 9 | 1520 | 109 | ||
| BC098506.1 | - | 9 - 10 | 1778 | 30014037 | 109 | |
| BC116304.1 | - | 8 - 9 | 1270 | 109 | ||
| BC116305.1 | - | 8 - 9 | 1270 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec23ip
Genomic Map