Assay Details
| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113373.2 | NP_001106844.2 | 4 - 5 | 1039 | 82 | |
| GenBank mRNA | AB099695.1 | - | 4 - 5 | 666 | 82 | |
| AK139360.1 | - | 4 - 5 | 1040 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shank2
Genomic Map