Assay Details
| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113373.2 | NP_001106844.2 | 3 - 4 | 921 | 68 | |
| GenBank mRNA | AB099695.1 | - | 3 - 4 | 548 | 68 | |
| AK139360.1 | - | 3 - 4 | 922 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shank2
Genomic Map