| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600003259.mm.1 |
TC0600000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003259.mm.1 |
NM_001171003.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003259.mm.1 |
NM_001171003.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003240.mm.1 |
TC0600000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003240.mm.1 |
NM_001171003.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003240.mm.1 |
NM_001171003.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003258.mm.1 |
TC0600000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003258.mm.1 |
NM_001171003.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003258.mm.1 |
NM_001171003.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003257.mm.1 |
TC0600000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003257.mm.1 |
NM_001171003.1 |
Refseq |