Assay Details
| Gene Symbol: Mgam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171003.1 | NP_001164474.1 | 35 - 36 | 4340 | 67 | |
| GenBank mRNA | AK031332.1 | - | 19 - 20 | 2344 | 67 | |
| EU937529.1 | - | 35 - 36 | 4340 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mgam
Genomic Map