Assay Details
| Gene Symbol: Mgam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171003.1 | NP_001164474.1 | 36 - 37 | 4427 | 88 | |
| GenBank mRNA | AK031332.1 | - | 20 - 21 | 2431 | 88 | |
| BC025509.1 | - | 2 - 3 | 127 | 88 | ||
| EU937529.1 | - | 36 - 37 | 4427 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgam
Genomic Map