Assay Details
| Gene Symbol: Mgam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171003.1 | NP_001164474.1 | 40 - 41 | 4859 | 106 | |
| GenBank mRNA | AK031332.1 | - | 24 - 25 | 2863 | 105 | |
| BC025509.1 | - | 6 - 7 | 559 | 106 | ||
| EU937529.1 | - | 40 - 41 | 4859 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgam
Genomic Map