Assay Details
| Gene Symbol: Mgam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171003.1 | NP_001164474.1 | 44 - 45 | 5271 | 55 | |
| GenBank mRNA | AK031332.1 | - | 28 - 29 | 3274 | 55 | |
| AK033548.1 | - | 4 - 5 | 410 | 55 | ||
| BC025509.1 | - | 10 - 11 | 971 | 55 | ||
| EU937529.1 | - | 44 - 45 | 5271 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgam
Genomic Map