| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600003210.mm.1 |
TC0600000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003210.mm.1 |
NM_001171003.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003210.mm.1 |
NM_001171003.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003211.mm.1 |
TC0600000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003211.mm.1 |
NM_001171003.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003211.mm.1 |
NM_001171003.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003212.mm.1 |
TC0600000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003212.mm.1 |
NM_001171003.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003212.mm.1 |
NM_001171003.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003213.mm.1 |
TC0600000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003213.mm.1 |
NM_001171003.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003213.mm.1 |
NM_001171003.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003214.mm.1 |
TC0600000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003214.mm.1 |
NM_001171003.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003214.mm.1 |
NM_001171003.1 |
Refseq |