Assay Details
| Gene Symbol: Mgam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171003.1 | NP_001164474.1 | 9 - 10 | 1186 | 82 | |
| GenBank mRNA | AY229885.1 | - | 1186 | 82 | ||
| EU937529.1 | - | 9 - 10 | 1186 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgam
Genomic Map