Assay Details
| Gene Symbol: Prl2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025532.3 | NP_079808.1 | 2 - 3 | 290 | 140 | |
| GenBank mRNA | AF525155.1 | - | 2 - 3 | 241 | 140 | |
| AK005411.1 | - | 2 - 3 | 291 | 140 | ||
| AK009863.1 | - | 2 - 3 | 290 | 139 | ||
| BC100329.1 | - | 2 - 3 | 280 | 30787440 | 140 | |
| BC111112.1 | - | 2 - 3 | 286 | 40050224 | 140 | |
| BC138199.1 | - | 2 - 3 | 283 | 8861218 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prl2b1
Genomic Map