Assay Details
| Gene Symbol: Prl2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025532.3 | NP_079808.1 | 4 - 5 | 570 | 75 | |
| GenBank mRNA | AF525155.1 | - | 4 - 5 | 521 | 75 | |
| AK005411.1 | - | 4 - 5 | 571 | 75 | ||
| BC100329.1 | - | 4 - 5 | 560 | 30787440 | 75 | |
| BC111112.1 | - | 4 - 5 | 566 | 40050224 | 75 | |
| BC138199.1 | - | 4 - 5 | 563 | 8861218 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028898 | BC100329.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028898 | NM_025532.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028898 | AY077703.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26368 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028898 | BC100329.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028898 | NM_025532.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028898 | AY077703.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902039 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07818532 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07818552 | - | - |
| Whole Mouse Genome G4122A | A_51_P413665 | BC138199.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413665 | BC100329.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413665 | BC111112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413665 | NM_025532.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413665 | AK005411.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P138998 | BC100329.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P138998 | BC111112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P138998 | NM_025532.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P138998 | AK005411.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P138998 | AY077703.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135267 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07818532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902080 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408377 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408376 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408375 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408378 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408374 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452165_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426730_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408372 | - | - |
| Mouse Genome 430 2.0 Array | 1452165_at | - | - |
| Mouse Genome 430 2.0 Array | 1452165_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426730_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426730_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66392_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66392_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300010484.mm.1 | NM_025532.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010484.mm.1 | NM_025532.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010484.mm.1 | TC1300001689.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010483.mm.1 | NM_025532.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010483.mm.1 | NM_025532.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010483.mm.1 | TC1300001689.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010482.mm.1 | NM_025532.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010482.mm.1 | NM_025532.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010482.mm.1 | TC1300001689.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452165_PM_at | - | - |
| Mouse Expression Set 430 | 1426730_a_at | - | - |
| Mouse Expression Set 430 | 1426730_PM_a_at | - | - |
| Mouse Expression Set 430 | 1452165_at | - | - |
| Murine Genome U74v2 Set | 160525_f_at | - | - |
| Murine Genome U74v2 Set | 160525_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452165_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426730_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426730_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452165_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452165_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452165_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426730_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426730_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 445938 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4850541 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4850541 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850541 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215934 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215934 | - | - |
| mouse-6 v1.1 (Illumina) | scl44173.5.22_179-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850541 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44173.5.22_179-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850541 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1215934 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215934 | - | - |

Prl2b1
Genomic Map