Assay Details
| Gene Symbol: Folr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252552.1 | NP_001239481.1 | 1 - 2 | 177 | 77 | |
| NM_001252554.1 | NP_001239483.1 | 1 - 2 | 177 | 77 | ||
| NM_008034.3 | NP_032060.2 | 1 - 2 | 177 | 77 | ||
| GenBank mRNA | AF096319.1 | - | 1 - 2 | 72 | 77 | |
| CJ079096.1 | - | 1 - 2 | 112 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF096319.1, NM_001252552.1, NM_001252554.1, NM_008034.3 transcripts.
Set Membership: |
|

Folr1
Genomic Map