Assay Details
| Gene Symbol: Ankrd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175134.4 | NP_780343.1 | 2 - 3 | 188 | 119 | |
| GenBank mRNA | AB076382.1 | - | 2 - 3 | 183 | 119 | |
| AK088304.1 | - | 2 - 3 | 149 | 119 | ||
| AK148049.1 | - | 2 - 3 | 149 | 119 | ||
| BC035553.1 | - | 2 - 3 | 189 | 119 | ||
| BC086659.1 | - | 2 - 3 | 155 | 30607272 | 119 | |
| BC094595.1 | - | 2 - 3 | 151 | 5707829 | 119 | |
| BQ715954.1 | - | 2 - 3 | 188 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB076382.1, AK088304.1, AK148049.1, BC086659.1, BC094595.1, NM_175134.4 transcripts.
Set Membership: |
|

Ankrd46
Genomic Map