Assay Details
| Gene Symbol: Piga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011081.2 | NP_035211.2 | 1 - 2 | 55 | 60 | |
| GenBank mRNA | AK039802.1 | - | 1 - 2 | 55 | 60 | |
| BY270710.1 | - | 1 - 2 | 50 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_011081.2 transcripts.
Set Membership: |
|

Piga
Genomic Map