Assay Details
| Gene Symbol: Gm2824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033069.1 | - | 1 - 2 | 78 | 83 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2335045 | AK083927.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425010 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906389 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940815 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126007 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51667 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15887 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43657 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132107 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2335045 | AK083927.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993733 | AK033069.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08311039 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08311082 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08311101 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08311060 | - | - |
| Whole Mouse Genome G4122A | A_51_P311642 | AK083927.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241834 | AK033069.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114631 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08311101 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08311039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12340443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11886750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12454752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11918560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12042240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30078963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12424103 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1459006_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1459006_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1459006_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100040529_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100040529_PM_at | - | - |
| Mouse Expression Set 430 | 1459006_PM_a_at | - | - |
| Mouse Expression Set 430 | 1459006_a_at | - | - |
| Murine Genome U74v2 Set | 136239_PM_at | - | - |
| Murine Genome U74v2 Set | 136239_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101940400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101940400 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232944 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101940400 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101940400 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232944 | - | - |

Gm2824
Genomic Map