Assay Details
| Gene Symbol: Tmbim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026617.3 | NP_080893.1 | 3 - 4 | 332 | 104 | |
| GenBank mRNA | AK006207.1 | - | 3 - 4 | 333 | 104 | |
| AK208947.1 | - | 1 - 2 | 63 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmbim4
Genomic Map