Assay Details
| Gene Symbol: Helb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080446.2 | NP_536694.2 | 7 - 8 | 2192 | 96 | |
| GenBank mRNA | AB048542.1 | - | 7 - 8 | 2117 | 96 | |
| AK131122.1 | - | 7 - 8 | 2214 | 96 | ||
| BC068140.1 | - | 7 - 8 | 2120 | 6843107 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Helb
Genomic Map