Assay Details
| Gene Symbol: Grip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277292.1 | NP_001264221.1 | 7 - 8 | 1062 | 62 | |
| NM_001277293.1 | NP_001264222.1 | 7 - 8 | 1062 | 62 | ||
| NM_001277294.1 | NP_001264223.1 | 7 - 8 | 919 | 62 | ||
| NM_028736.2 | NP_083012.1 | 7 - 8 | 919 | 62 | ||
| NM_130891.2 | NP_570961.1 | 6 - 7 | 838 | 62 | ||
| NM_133442.2 | NP_597699.1 | 6 - 7 | 981 | 62 | ||
| GenBank mRNA | AB051560.1 | - | 6 - 7 | 715 | 62 | |
| AB051561.1 | - | 7 - 8 | 796 | 62 | ||
| AB051562.1 | - | 6 - 7 | 701 | 62 | ||
| AK029845.1 | - | 7 - 8 | 717 | 62 | ||
| AK039673.1 | - | 6 - 7 | 723 | 62 | ||
| AK220553.1 | - | 7 - 8 | 776 | 62 | ||
| BC067398.1 | - | 7 - 8 | 919 | 30096759 | 62 | |
| BC072632.1 | - | 7 - 8 | 1062 | 6843802 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grip1
Genomic Map