Assay Details
| Gene Symbol: Ankhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175375.3 | NP_780584.2 | 15 - 16 | 3149 | 79 | |
| GenBank mRNA | AK029322.1 | - | 8 - 9 | 1564 | 79 | |
| AK131166.1 | - | 15 - 16 | 3012 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankhd1
Genomic Map