Assay Details
| Gene Symbol: Gpr141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181754.4 | NP_861419.3 | 3 - 4 | 304 | 71 | |
| GenBank mRNA | AK154688.1 | - | 3 - 4 | 305 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154688.1, NM_181754.4 transcripts.
Set Membership: |
|

Gpr141
Genomic Map