Assay Details
| Gene Symbol: Asxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167777.1 | NP_001161249.1 | 11 - 12 | 1213 | 146 | |
| GenBank mRNA | AK082659.1 | - | 11 - 12 | 1213 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asxl3
Genomic Map