Assay Details
| Gene Symbol: Asxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167777.1 | NP_001161249.1 | 1 - 2 | 80 | 74 | |
| GenBank mRNA | AK082659.1 | - | 1 - 2 | 80 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK082659.1, NM_001167777.1 transcripts.
Set Membership: |
|

Asxl3
Genomic Map