Assay Details
| Gene Symbol: 9330182O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256056.1 | NP_001242985.1 | 2 - 3 | 394 | 117 | |
| GenBank mRNA | AK034361.1 | - | 2 - 3 | 396 | 117 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 33251 |
Gm33251 | -13.1kb | Sense | Chr. 15 - 40045191 - 40128982 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 33301 |
Gm33301 | 17.6kb | Sense | Chr. 15 - 40162603 - 40168076 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0700003034.rn
Set Membership: |
|

9330182O14Rik
Genomic Map