Assay Details
| Gene Symbol: Gm36642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086666.1 | - | 5 - 6 | 1356 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5468 | Gm5468 |
100040583, EG432939 |
-76.5kb | Anti-sense | Chr. 15 - 25414192 - 25452418 on NCBI Build GRCm38 |
| Downstream | myosin X | Myo10 |
AW048724, D15Ertd600e, mKIAA0799 |
92.3kb | Anti-sense | Chr. 15 - 25622550 - 25813673 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm36642
Genomic Map