Assay Details
| Gene Symbol: Gm13848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK085570.1 | - | 1 - 2 | 199 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | podocalyxin-like | Podxl |
AW121214, Ly102, PC, PCLP-1, Pclp1 |
-99.9kb | Sense | Chr. 6 - 31519493 - 31563937 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700012A03 gene | 1700012A03Rik | AI429084 | 384.2kb | Anti-sense | Chr. 6 - 32050285 - 32058915 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm13848
Genomic Map