Assay Details
| Gene Symbol: Gm38403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110482.1 | 1 - 2 | 105 | 90 | ||
| GenBank mRNA | AK082258.1 | - | 1 - 2 | 106 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2 | Dyrk2 | 1810038L18Rik | -36.9kb | Sense | Chr. 10 - 118855602 - 118889246 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 33677 |
Gm33677 | 101.1kb | Sense | Chr. 10 - 119028287 - 119053655 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm38403
Genomic Map