Assay Details
| Gene Symbol: Gm16291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045788.1 | 2 - 3 | 580 | 105 | ||
| GenBank mRNA | AK038507.1 | - | 2 - 3 | 581 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dihydropyrimidinase | Dpys |
1200017I10Rik, 1300004I01Rik, DHP, DHPase |
Anti-sense | Chr. 15 - 39768485 - 39857470 on NCBI Build GRCm38 | |
| Downstream | low density lipoprotein-related protein 12 | Lrp12 |
AI848829, C820005L12Rik, LRP-12 |
58.2kb | Anti-sense | Chr. 15 - 39870602 - 39944009 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16291
Genomic Map