Assay Details
| Gene Symbol: BC024386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC024386.1 | - | 2 - 3 | 308 | 74 | |
| BC100589.1 | - | 2 - 3 | 347 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

BC024386
Genomic Map