Assay Details
| Gene Symbol: Gm35147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035726.1 | - | 1 - 2 | 175 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 39102 |
Gm39102 | -42.1kb | Anti-sense | Chr. 7 - 132420030 - 132423713 on NCBI Build GRCm38 | |
| Downstream | ornithine aminotransferase | Oat | AI194874 | 90.8kb | Sense | Chr. 7 - 132557475 - 132576398 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm35147
Genomic Map