Assay Details
| Gene Symbol: Zc3h10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134003.1 | NP_598764.1 | 1 - 2 | 55 | 62 | |
| GenBank mRNA | AK031312.1 | - | 86 | 62 | ||
| AK050662.1 | - | 1 - 2 | 49 | 62 | ||
| BC022677.1 | - | 1 - 2 | 55 | 4239511 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050662.1, BC022677.1, NM_134003.1 transcripts.
Set Membership: |
|

Zc3h10
Genomic Map