Assay Details
| Gene Symbol: Btn2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289614.1 | NP_001276543.1 | 3 - 4 | 696 | 79 | |
| NM_001289615.1 | NP_001276544.1 | 3 - 4 | 637 | 79 | ||
| NM_175938.3 | NP_787952.2 | 3 - 4 | 696 | 79 | ||
| GenBank mRNA | AK051061.1 | - | 3 - 4 | 683 | 79 | |
| AY177686.1 | - | 3 - 4 | 679 | 79 | ||
| BC139760.1 | - | 3 - 4 | 523 | 40058494 | 79 | |
| BC141341.1 | - | 3 - 4 | 669 | 9055953 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Btn2a2
Genomic Map