Assay Details
| Gene Symbol: Gm11377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK085428.1 | - | 331 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box Q1 | Foxq1 | HFH-1, Hfh1, Hfh1l, sa | -5.2kb | Sense | Chr. 13 - 31558170 - 31560976 on NCBI Build GRCm38 | 
| Downstream | forkhead box F2 | Foxf2 | FREAC2, Fkh20, LUN | 52.3kb | Sense | Chr. 13 - 31625816 - 31631406 on NCBI Build GRCm38 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  3' Most  Mus musculus  Probe spans exons | 
 Back To Top
 Back To Top
 Gm11377
Gm11377 Genomic Map
Genomic Map Back To Top
 Back To Top