Assay Details
| Gene Symbol: Prl7b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029355.2 | NP_083631.1 | 3 - 4 | 311 | 120 | |
| GenBank mRNA | AF525156.1 | - | 3 - 4 | 311 | 120 | |
| AK005458.1 | - | 3 - 4 | 312 | 120 | ||
| BC116772.1 | - | 3 - 4 | 288 | 40126091 | 120 | |
| BC119183.1 | - | 3 - 4 | 288 | 8733932 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prl7b1
Genomic Map