Assay Details
| Gene Symbol: Gm15245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079668.1 | - | 5 - 6 | 538 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | holocytochrome c synthetase | Hccs | -152.6kb | Anti-sense | Chr. X - 169311530 - 169320343 on NCBI Build GRCm38 | |
| Downstream | midline 1 | Mid1 |
61B3-R, DXHXS1141, Fxy, Trim18 |
209.8kb | Sense | Chr. Y - 90718657 - 90765072 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15245
Genomic Map