| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800008008.mm.1 |
NM_001146292.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008008.mm.1 |
NM_001146292.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008008.mm.1 |
NM_001146294.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008008.mm.1 |
NM_001146294.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008008.mm.1 |
TC1800001088.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800008009.mm.1 |
NM_001146292.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008009.mm.1 |
NM_001146292.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008009.mm.1 |
NM_001146293.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008009.mm.1 |
NM_001146293.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008009.mm.1 |
NM_001146294.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008009.mm.1 |
NM_001146294.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008009.mm.1 |
NM_133195.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008009.mm.1 |
NM_133195.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008009.mm.1 |
TC1800001088.mm.1 |
MTA TC |