Assay Details
| Gene Symbol: Cdk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081058.2 | NP_001074527.1 | 11 - 12 | 3471 | 123 | |
| NM_027118.1 | NP_081394.1 | 11 - 12 | 3471 | 123 | ||
| GenBank mRNA | AK035493.1 | - | 11 - 12 | 2142 | 123 | |
| BC051093.1 | - | 4 - 5 | 355 | 123 | ||
| BC076619.1 | - | 12 - 13 | 1926 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk13
Genomic Map