Assay Details
| Gene Symbol: Cdk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081058.2 | NP_001074527.1 | 4 - 5 | 2630 | 153 | |
| NM_027118.1 | NP_081394.1 | 4 - 5 | 2630 | 153 | ||
| GenBank mRNA | AK035493.1 | - | 4 - 5 | 1301 | 153 | |
| AK173265.1 | - | 4 - 5 | 2630 | 153 | ||
| BC076619.1 | - | 5 - 6 | 1085 | 153 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk13
Genomic Map