Assay Details
| Gene Symbol: Cdk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081058.2 | NP_001074527.1 | 8 - 9 | 3145 | 102 | |
| NM_027118.1 | NP_081394.1 | 8 - 9 | 3145 | 102 | ||
| GenBank mRNA | AK035493.1 | - | 8 - 9 | 1816 | 102 | |
| AK173265.1 | - | 8 - 9 | 3145 | 102 | ||
| BC076619.1 | - | 9 - 10 | 1600 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk13
Genomic Map