Assay Details
| Gene Symbol: Gm16090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090335.1 | - | 2 - 3 | 985 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | desmoglein 3 | Dsg3 |
bal, balding |
-56.7kb | Anti-sense | Chr. 18 - 20510304 - 20541310 on NCBI Build GRCm38 |
| Downstream | transthyretin | Ttr |
AA408768, AI787086, D17860, prealbumin |
66.2kb | Anti-sense | Chr. 18 - 20665250 - 20674326 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16090
Genomic Map