Assay Details
| Gene Symbol: 4930528H21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019707.1 | - | 1 - 2 | 130 | 97 | |
| BY715486.1 | - | 1 - 2 | 130 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
guanine nucleotide binding protein (G protein), gamma 11 |
Gng11 | 0610037B21Rik | -36.4kb | Anti-sense | Chr. 6 - 4003987 - 4008445 on NCBI Build GRCm38 |
| Downstream | blocked early in transport 1 homolog (S. cerevisiae) | Bet1 |
AW555236, Bet-1 |
29.3kb | Sense | Chr. 6 - 4076904 - 4086995 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930528H21Rik
Genomic Map