Assay Details
| Gene Symbol: 9330158H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015589.1 | 2 - 3 | 370 | 69 | ||
| GenBank mRNA | AK034128.1 | - | 2 - 3 | 371 | 69 | |
| AK043142.1 | - | 427 | 69 | |||
| AK139997.1 | - | 187 | 69 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cholinergic receptor, muscarinic 2, cardiac |
Chrm2 |
AChR-M2, Chrm-2, M2 |
Anti-sense | Chr. 6 - 36387950 - 36528638 on NCBI Build GRCm38 | |
| Downstream | microRNA 490 | Mir490 |
Mirn490, mir-490, mmu-mir-490 |
33.8kb | Anti-sense | Chr. 6 - 36421742 - 36421825 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139997.1 transcripts.
Set Membership: |
|

9330158H04Rik
Genomic Map